About this item
Flow adapters are available for each of the column Ø to allow optimal bed heights for each application, as are thermostatic jackets for precise temperature control.
The basic C columns are of fixed volume but can be upgraded by adding flow adapters (which keep bed height constant and protect from insoluble particles) and a thermostatic jacket. Three adjustable flow adapters are available for the C column series (AC 10, AC 16, and AC 26), allowing the choice of bed height to suit the application.
Resistant to aqueous solutions and solvents used in liquid chromatography. Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.
Maximum pressure: 1 bar (0.1 MPa ) (14.5 psi)
Maximum temperature: 4 to 60 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements