Whatman™ Grade 556 Blotting Papers, Whatman products (Cytiva)
10034-634
: Cytiva
Product available on GSA Advantage®
- Description:Grade 556 Drypads
- Dimensions:37 x 100 mm
- Cat. No.:10034-634
- Supplier no.:10310992
Absorbent paper strips made from cellulose for the removal of excess liquids, e.g., from microscope specimens.