Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Methanol ≥99%

Methanol ≥99%

Methanol ≥99%
Supplier:  BeanTown Chemical
Certificates
Methanol ≥99%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000 UN No: UN1230; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3OH; MW: 32.04 Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F) Density (g/mL): 0.791; Refractive Index: 1.329

  • RTECS: PC1400000
  • Refractive index: 1.329
  • Liquid
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Boiling Pt:64.7 °C
  • Melting Pt:–98 °C
  • Density:0.791 g/mL
  • Flash Pt:9.7 °C (49.5 °F)
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    99%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $23.90
    $23.90
    Each (500ml)
       $23.90

    Supplier Number: 223840-500ML
  • 1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $30.20
    $30.20
    Each (1l)
       $30.20

    Supplier Number: 223840-1L
  • 2.5 L
    Log in for availability
    Log in for availability
    Each (2,500ml)
    Each (2,500ml) - $58.10
    $58.10
    Each (2,500ml)
       $58.10

    Supplier Number: 223840-2.5L

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.