Order Entry
Puerto Rico
ContactUsLinkComponent
Iridium, Plasma Standard Solution, Ir 1000 µg/ml,
Iridium, Plasma Standard Solution, Ir 1000 µg/ml,
Supplier:  BeanTown Chemical
Certificates
Iridium, Plasma Standard Solution, Ir 1000 µg/ml,
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00011062
  • UN No.:
    1789
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together