Order Entry
Puerto Rico
ContactUsLinkComponent
 
Cobalt, Plasma Standard Solution, Co 10000 µg/ml
 
Cobalt, Plasma Standard Solution, Co 10000 µg/ml

 

  • MDL No.:
    MFCD00010935
  • UN No.:
    3264
  • GHS signal:
    Danger