Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Zirconium Standards
Zirconium, Plasma Standard Solution, Zr 1000 µg/ml
Zirconium, Plasma Standard Solution, Zr 1000 µg/ml
Zirconium, Plasma Standard Solution, Zr 1000 µg/ml
Supplier:
BeanTown Chemical
Certificates
Expand image
Zirconium, Plasma Standard Solution, Zr 1000 µg/ml
Supplier:
BeanTown Chemical
About this item
Specifications
MDL No.:
MFCD00011303
UN No.:
1789
GHS signal:
Danger
Specifications
Related Information
Documentation
SDS
ghs_219075
Documentation