Order Entry
Puerto Rico
ContactUsLinkComponent
Platinum, AAS Standard Solution, Pt 1000 µg/ml
Platinum, AAS Standard Solution, Pt 1000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Platinum, AAS Standard Solution, Pt 1000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00011179
  • UN No.:
    1789
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together