Order Entry
Puerto Rico
ContactUsLinkComponent
Lanthanum(III) chloride releasing agent solution
Lanthanum(III) chloride releasing agent solution
Catalog # BT216610-500ML
Supplier:  BeanTown Chemical
CAS Number:  
Lanthanum(III) chloride releasing agent solution
Catalog # BT216610-500ML
Supplier:  BeanTown Chemical
Supplier Number:  216610-500ML
CAS Number:  

Specifications

  • Size:
    500 ml
  • Cat. No.:
    BT216610-500ML
  • Supplier no.:
    216610-500ML

Specifications

About this item

 :
 :245.26 g/mol
EINECS:231-595-7
ADR:8,III
CAS Number:10099-58-8
Formula:Cl₃La
MDL Number:MFCD00011068