Order Entry
Puerto Rico
ContactUsLinkComponent
Titanium, Plasma Standard Solution, Ti 1000 µg/ml
Titanium, Plasma Standard Solution, Ti 1000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Titanium, Plasma Standard Solution, Ti 1000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00011264
  • UN No.:
    3264
  • GHS signal:
    Danger

Specifications

Related Information