Sodium hydroxide 10.0 N in water volumetric solution
BT214540-1L
:
- Pack type:HDPE bottle
- Size:1 L
- Cat. No.:BT214540-1L
- Supplier no.:214540-1L
:
:40 g/mol
:105…140 °C
:–12–10 °C
:1.327 g/mL
:215-185-5
:1824
:8,II
:1310-73-2
:NaOH
:MFCD00003548