Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lithium tetrachlorocuprate
Lithium tetrachlorocuprate 0.1 M in THF
Lithium tetrachlorocuprate 0.1 M in THF
Lithium tetrachlorocuprate 0.1 M in THF
Supplier:
BeanTown Chemical
Certificates
Expand image
Lithium tetrachlorocuprate 0.1 M in THF
Supplier:
BeanTown Chemical
About this item
CAS Number:
15489-27-7
:
Formula:
Cl₄CuLi₂
MDL Number:
MFCD00011081
:
219.24 g/mol
Density:
0.91 g/mL
ADR:
3,II
Related Information
Documentation
SDS
ghs_212710.pdf
Documentation