Zirconium, Plasma Standard Solution, Zr 10000 µg/ml
Supplier: BeanTown Chemical
Certificates
About this item
EC No: 231-595-7
Haz class: 8
Packing group: III
GHS symbol: GHS05
Specifications
- MDL No.:MFCD00011303
- UN No.:1789
- GHS signal:Danger
EC No: 231-595-7
Haz class: 8
Packing group: III
GHS symbol: GHS05