Sodium hydroxide 1.0 N in water volumetric solution
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:105…140 °C
Melting Pt:–12–10 °C
Density:1.04 g/mL
EINECS:215-185-5
UN:3266
ADR:8,III