Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Titanium(III) sulfate
Titanium(III) sulfate 20% (w/w) in sulfuric acid (4%)
Titanium(III) sulfate 20% (w/w) in sulfuric acid (4%)
Titanium(III) sulfate 20% (w/w) in sulfuric acid (4%)
Supplier:
BeanTown Chemical
Certificates
Expand image
Titanium(III) sulfate 20% (w/w) in sulfuric acid (4%)
Supplier:
BeanTown Chemical
About this item
CAS Number:
10343-61-0
:
Formula:
O₁₂S₃Ti₂
MDL Number:
MFCD00036317
:
383.902 g/mol
Density:
1.456 g/mL
ADR:
8,III
Related Information
Documentation
SDS
ghs_145480.pdf
Documentation