$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Ammonia solution
Ammonia solution 25% ≥99.99% (trace metals basis)
Ammonia solution 25% ≥99.99% (trace metals basis)
:
BeanTown Chemical
Ammonia solution 25% ≥99.99% (trace metals basis)
:
BeanTown Chemical
:
1336-21-6
:
7664-41-7
:
NH₃
:
MFCD00066650
:
35.05 g/mol
:
0.9 g/mL
:
215-647-6
:
2672
:
8,III
Assay:
99.99%
ghs_142850.pdf