Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Beryllium Standards
Beryllium, Plasma Standard Solution, Be 1000 µg/ml
Beryllium, Plasma Standard Solution, Be 1000 µg/ml
:
BeanTown Chemical
Beryllium, Plasma Standard Solution, Be 1000 µg/ml
:
BeanTown Chemical
MDL No.:
MFCD00134032
UN No.:
3264
GHS signal:
Danger
ghs_141095