Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonia solution
Ammonia solution 5.0 N in water
Ammonia solution 5.0 N in water
Ammonia solution 5.0 N in water
Supplier:
BeanTown Chemical
Certificates
Expand image
Ammonia solution 5.0 N in water
Supplier:
BeanTown Chemical
Certificates
About this item
CAS Number:
1336-21-6
:
7664-41-7
Formula:
NH₃
MDL Number:
MFCD00066650
:
35.05 g/mol
Density:
0.9 g/mL
EINECS:
215-647-6
UN:
2672
ADR:
8,III
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_139710.pdf
Documentation