Diethyl ether ≥99.0% stabilized ACS
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Boiling Pt:34.6 °C
Melting Pt:–116 °C
Density:0.706 g/mL
Flash Pt:–40 °C (–40 °F)
EINECS:200-467-2
UN:1155
ADR:3,I
Specification Test Results
- Assay:99.0% min.
- Color (APHA):≤10
- Residue after evaporation:≤10ppm
- Titrable acid:≤0.0002meq/g
- Carbonyl (as HCHO):≤0.001%
- Peroxide (as H2O2):≤1ppm
- Stabilizer (BHT):5ppm
- Ethanol:≤0.05%
- Water:≤0.03%