Order Entry
Puerto Rico
ContactUsLinkComponent
Ruthenium, Plasma Standard Solution, Ru 1000 µg/ml
Ruthenium, Plasma Standard Solution, Ru 1000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Ruthenium, Plasma Standard Solution, Ru 1000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00011207
  • UN No.:
    1789
  • GHS signal:
    Danger

Specifications

Related Information