Ethanolamine ≥98%
About this item
CAS: 141-43-5; EC No: 205-483-3; MDL No: MFCD00008183; RTECS: KJ5775000 UN No: UN2491; Haz Class: 8; Packing Group: III Liquid; Linear Formula: HOCH2CH2NH2; Molecular Formula: C2H7NO; MW: 61.08 Melting Point: 10-11°; Boiling Point: 170°; Flash point: 86°C (187°F) Density (g/mL): 1.012; Refractive Index: 1.454 Air Sensitive , Hygroscopic
- RTECS: KJ5775000
- Refractive index: 1.454
- Air Sensitive, Hygroscopic
- Liquid
- CAS Number:141-43-5
- Formula:NH₂CH₂CH₂OH
- MDL Number:MFCD00008183
- Molecular Weight:61.08 g/mol
- Boiling Pt:170 °C
- Melting Pt:10…11 °C
- Density:1.012 g/mL
- Flash Pt:86 °C (187 °F)
- EINECS:205-483-3
- UN:2491
- ADR:8,III
2 Options Available Below
Product Details & Documents
CAS: 141-43-5; EC No: 205-483-3; MDL No: MFCD00008183; RTECS: KJ5775000 UN No: UN2491; Haz Class: 8; Packing Group: III Liquid; Linear Formula: HOCH2CH2NH2; Molecular Formula: C2H7NO; MW: 61.08 Melting Point: 10-11°; Boiling Point: 170°; Flash point: 86°C (187°F) Density (g/mL): 1.012; Refractive Index: 1.454 Air Sensitive , Hygroscopic
- RTECS: KJ5775000
- Refractive index: 1.454
- Air Sensitive, Hygroscopic
- Liquid
- CAS Number:141-43-5
- Formula:NH₂CH₂CH₂OH
- MDL Number:MFCD00008183
- Molecular Weight:61.08 g/mol
- Boiling Pt:170 °C
- Melting Pt:10…11 °C
- Density:1.012 g/mL
- Flash Pt:86 °C (187 °F)
- EINECS:205-483-3
- UN:2491
- ADR:8,III
Specifications for Product Family
Specification Test Results
- Assay:98%
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



