Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Tin(II) bromide ≥99%

Tin(II) bromide ≥99%
Supplier:  BeanTown Chemical
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 10031-24-0; EC No: 233-087-0; MDL No: MFCD00011239 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: SnBr2; MW: 278.51 Melting Point: 215°; Boiling Point: 623° Density (g/mL): 5.12 Air Sensitive, Light Sensitive, Moisture Sensitive

  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Powder
Synonyms:  Tin dibromide
  • CAS Number:10031-24-0
  • Formula:Br₂Sn
  • MDL Number:MFCD00011239
  • Molecular Weight:278.52 g/mol
  • Boiling Pt:623 °C
  • Melting Pt:215 °C
  • Density:5.12 g/mL
  • EINECS:233-087-0
  • UN:3260
  • ADR:8,III
Specifications for Product Family

Specification Test Results

  • Assay:
    99%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $21.30
    $21.30
    Each (5g)
       $21.30

    Supplier Number: 129015-5G
  • 25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $94.60
    $94.60
    Each (25g)
       $94.60

    Supplier Number: 129015-25G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.