Order Entry
Puerto Rico
ContactUsLinkComponent
Copper, AAS Standard Solution, Cu 1000 µg/ml
Copper, AAS Standard Solution, Cu 1000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Copper, AAS Standard Solution, Cu 1000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00010965
  • UN No.:
    3264
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together