Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dichloromethane
Dichloromethane ≥99.5% stabilized ACS
Dichloromethane ≥99.5% stabilized ACS
:
BeanTown Chemical
Dichloromethane ≥99.5% stabilized ACS
:
BeanTown Chemical
:
75-09-2
:
:
CH₂Cl₂
:
MFCD00000881
:
84.93 g/mol
:
39…40 °C
:
–97 °C
:
1.325 g/mL
:
200-838-9
:
1593
:
6.1,III
Assay:
99.5% min.
ghs_123885