Order Entry
Puerto Rico
ContactUsLinkComponent
 
Copper(II) tetrafluoroborate 45% (w/w) in water
Catalog # BT123690-1KG
Supplier:  BeanTown Chemical
CAS Number:  
Copper(II) tetrafluoroborate 45% (w/w) in water
Catalog # BT123690-1KG
Supplier:  BeanTown Chemical
Supplier Number:  123690-1KG
CAS Number:  

Specifications

  • Size:
    1 kg
  • Cat. No.:
    BT123690-1KG
  • Supplier no.:
    123690-1KG

Specifications

About this item

 :
 :237.15 g/mol
EINECS:253-959-4
ADR:8,II
CAS Number:38465-60-0
Formula:B₂CuF₈
MDL Number:MFCD00016054