Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10065-754
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rabbit
- Western Blot:Yes
- Size:1 mg
- Epitope:Heavy and Light chain
- Storage Temperature:Store at 4°C
- Tested Applications:Plate-Based Binding Assays
- Cat. No.:10065-754
- Supplier no.:HRP-114
Specifications
About this item
Goat Polyclonal Secondary Antibody, reacts with Rabbit IgG.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit