Order Entry
Puerto Rico
ContactUsLinkComponent
Easy Na Sodium Analyzer
Easy Na Sodium Analyzer
Catalog # 10803-512
Supplier:  Mettler Toledo
cUL Listed
CSA Certified
UL Listed
undefined
Easy Na Sodium Analyzer
Catalog # 10803-512
Supplier:  Mettler Toledo
Supplier Number:  30060051
cUL Listed
CSA Certified
UL Listed

Specifications

  • Communication Interface:
    USB A (Printer or Balance), USB B (EasyDirect PC software)
  • Description:
    Easy Na sodium analyzer
  • Measurement Method:
    Multiple standard addition
  • Electrical:
    100 - 240 V, ±10%, 50/60 Hz, 1.25 A
  • Overall Dimensions:
    170×220×350 mm; 2650 g
  • User Interface:
    iTitrate
  • Cat. No.:
    10803-512
  • Supplier no.:
    30060051

Specifications

About this item

The Easy Na Sodium analyzer from METTLER TOLEDO determines the sodium content in food and other products simply and quickly.

  • Easy to use
  • Smartphone-style apps design
  • One LongClick™ needed
  • Save money
  • No calibration necessary
  • Simple result data management

Reduce sample preparation time and effort by using safer and less expensive chemicals while, specifically and exclusively measuring sodium ions.

Replace the silver nitrate (AgNO₃) titrant normally used for chloride determination with the less expensive and safer sodium reagents. Due to the standard addition technique procedure, no calibration is necessary. Just carry out the automated sensor conditioning and immediately start the analysis. Connect your sodium analyzer to EasyDirect™ Titration Software or to a USB-P25 dot-matrix printer.

Simply add a sample and ionic strength adjustment (ISA) solution, and start the measurement. Smartphone-style apps design is used to controls all major functions. One LongClick™ is all that is necessary to start the last analysis run.

Certifications: UL listed and CSA certified. IEC and EN certified.
Ordering information: Standard delivery–sodium analyzer with analyzer head, EasyStir and magnetic stir bar, 10 ml burette, bottle head for sodium standard, DX222-Na Sodium ISE, DX205-SC reference electrode and cable, tubing set, 110 V power adapter, EasyDirect data transfer software (trial), and Application Brochure.