- Description:ST Infinity H&E Staining System
- Cat. No.:10015-090
- Supplier no.:3801698
All stains and reagents are optimized for use with Leica ST5010 and Leica ST5020, resulting in high-quality H&E stains and consistent quality from the first stained slide to the last.
- Consistently high quality
- Enables high throughput of slides before stains expire
- Expediency allows all components to be loaded simultaneously
This system enables a high throughput of slides before the stains and reagents expire. The time saving, as all stains and reagents exhaust at the same rate, allows all components of the kit to be loaded on and off the instrument at the same time.