To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Neurogenin 3
- Antigen Symbol:NEUROG3
- Host:Multiple
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:1 Set
- Environmentally Preferable:
- Gene ID:50674
- Antigen Synonyms:NGN|bHLHa7|Atoh5|NGN1|Neurogenic basic-helix-loop-helix- protein|NGN-3|ngn3|Neurogenic differentiation factor 3|NEUROG1|NeuroD3|NEUROD3|Math4B
- Cat. No.:10540-588
- Supplier no.:H00050674-PW2
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-NEUROG3 (300 ?g); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-NEUROG3 (50 ?l)