Anti-PRKAG2 Polyclonal Antibody Pair
10541-832
: Abnova
- Antigen Name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 2
- Antigen Symbol:PRKAG2
- Clonality:Polyclonal
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Gene ID:51422
- Antigen Synonyms:CMH6|AAKG|AAKG2|WPWS|H91620p
- Cat. No.:10541-832
- Supplier no.:H00051422-PW1
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-PRKAG2 (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-PRKAG2 (50 µl)
Type:
Antigen: PRKAG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human