To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:CACYBP + TBL1X
- Host:Multiple
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Gene ID:27101|6907
- Cat. No.:10536-764
- Supplier no.:DI0621
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CACYBP protein, and the other against the TBL1X protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CACYBP Rabbit purified Polyclonal Antibody (20 µg) 2. TBL1X Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CACYBP + TBL1X
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype: IgG1
Reactivity: