Anti-PRKAB1 Antibody Pair
Catalog # 10541-830
Supplier: Abnova
Specifications
- Antigen Name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta
- Antigen Symbol:PRKAB1
- Host:Multiple
- Reactivity:Human
- Gene ID:5564
- Antigen Synonyms:kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
- Cat. No.:10541-830
- Supplier no.:H00005564-AP51
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human PRKAB1.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-PRKAB1 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-PRKAB1 (50 µg);
Type:
Antigen: PRKAB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human