ContactUsLinkComponent
Resprep® Resin SPE Disks, Restek
Resprep® Resin SPE Disks, Restek
Catalog # 10015-396
Supplier:  Restek
undefined
Resprep® Resin SPE Disks, Restek
Catalog # 10015-396
Supplier:  Restek
Supplier Number:  26023
Specifications
  • Description:
    Resprep® Resin SPE Disks, 47 mm I.D.
  • Cat. No.:
    10015-396
  • Supplier no.:
    26023
Specifications
About this item
These 47 mm glass fiber disks are embedded with styrene/DVB resin.
Resprep® resin SPE disks are designed for use with chlorinated, benzidine-containing, or nitrogen-containing pesticides.
Certifications: Disks meet requirements of EPA methods 515.2 and 553.