ContactUsLinkComponent
Resprep® Sample Delivery System, Restek
Resprep® Sample Delivery System, Restek
  10803-402
 :  Restek
undefined
Resprep® Sample Delivery System, Restek
  10803-402
 :  Restek
 :  26250
  • Description:
    Resprep® Sample Delivery System
  • Cat. No.:
    10803-402
  • Supplier no.:
    26250
Compatible with Resprep® 1 ml, 3 ml, 6 ml, and 15 ml SPE cartridges and the Diskcover-47 extraction disk holder.
  • Colored bands for easy identification
  • Compatible with SPE cartridges of different sizes
  • Prevents any contaminants leaching from system
The delivery system features six PTFE transfer lines (1/8" O.D.×1/16" I.D.×36" L). Each line is banded with a different color for easy sample identification. Specified in EPA drinking water methods. Tested to pH of 1 to ensure no contaminants leach from system.
 : Use the Resprep® sample delivery system to transfer large volumes of low-viscosity samples directly from a bottle to a solid phase extraction cartridge, or to a disk on a vacuum manifold system for extraction or cleanup.