About this item
Large Stainless Steel Pressure Filter Holders are offered in various sizes for batch type laboratory filtration using disc media.
Standard Holders are used in-line to filter gases and liquids such as culture media, solvents, photoresists. Sanitary Holders are threadless and can be disassembled for thorough cleaning. Fittings are 1.5" with tri-clamp. Use in-line to ultraclean or sterilize gases or liquids such as blood fractions, pure water and pharmaceutical applications.
The holder features a PTFE coated photoetched stainless steel filter support screen for autoclaving with filter in place. Unit comes standard with silicone o-ring. Other materials are available.
Units for filter diameters of 90 mm, 142 mm, and 293 mm are available in both Standard and Sanitary models. Optimal clean holder design minimizes resistance and maximizes flow rate.
Specifications
- Catalog No:
- 101105-054
- 101105-022
- 101105-722
- 101104-948
- 101105-852
- 101105-152
- 101104-602
- 101104-984
- 101105-008
- 101105-148
- 101105-150
- Max. Differential Pressure:
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- 42 psi
- Max. Inlet Pressure:
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi
- 99 psi