Glass Coverslips, 30mm, Bioptechs Inc.®
Catalog # 10199-874
Supplier: BIOPTECHS INC.
Specifications
- Ø:30 mm
- Numbering:#1.5
- Description:Glass cover slips, round
- Cat. No.:10199-874
- Supplier no.:3013130319
Specifications
About this item
Low alkaline, German Desag 263 glass coverslips are optimized for cell culture growth
Coverslips feature a 30mm diameter and #1.5 thickness.
Ordering information: Pack includes 100 coverslips.