Stage Spacer Kit, Bioptechs Inc.®
Catalog # 10199-812
Supplier: BIOPTECHS INC.
Specifications
- Description:Stage Spacer Kit, 9 mm
- Cat. No.:10199-812
- Supplier no.:191903
Specifications
About this item
This stage spacer kit is designed to raise the height of an inverted microscope stage.
Certifications: CE certified.
Delivery: Kit includes four 5×25 mm allen bolts and four 9 mm spacers.