2-Mercaptoethanol ≥99%, Molecular Biology Grade, Calbiochem®
Catalog # 80501-154
Supplier: MilliporeSigma
CAS Number:
Specifications
- Size:250 ml
- Cat. No.:80501-154
- Supplier no.:444203-250ML
Specifications
Specification Test Results
- Purity:≥99%
- DNases, proteases, RNases:none detected.
- Absorbance (260 nm):≤1.50
- Absorbance (280 nm):≤0.31
Specification Test Results
About this item
:
ADR:6.1,II
:78.14 g/mol
UN:2966
CAS Number:60-24-2
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890