Order Entry
Puerto Rico
ContactUsLinkComponent
FCS2®/FCS3® 30-Piece Gasket Set, Bioptechs Inc.®
FCS2®/FCS3® 30-Piece Gasket Set, Bioptechs Inc.®
Catalog # 10200-066
Supplier:  BIOPTECHS INC.
undefined
FCS2®/FCS3® 30-Piece Gasket Set, Bioptechs Inc.®
Catalog # 10200-066
Supplier:  BIOPTECHS INC.
Supplier Number:  06031920719

Specifications

  • Description:
    FCS2®/FCS3® 30-Piece Gasket Set
  • Material:
    Silicone
  • Cat. No.:
    10200-066
  • Supplier no.:
    06031920719

Specifications

About this item

A 30-piece assortment of round gaskets, featuring 14×22 mm and 30 mm round internal cutouts plus blanks of each thickness.

  • Unlimited options for internal geometry
  • Creates user definable flow characteristics
  • Enables near laminar flow

Designed for use with FCS2® or FCS3® chambers.

Certifications: CE certified.