n-Decane
Product Details & Documents
Reference Material for Flash Point Certified by The Japan Petroleum Institute
CAS Number: 124-18-5
MDL Number: MFCD00008954
Molecular Formula: C10H22
Molecular Weight: 142.29
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: Decane
- CAS Number:124-18-5
- Formula:H₃C(CH₂)₈CH₃
- MDL Number:MFCD00008954
- Molecular Weight:142.28 g/mol
- UN:2247
- ADR:3,III
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Size:250 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



