Reference Material of Gasoline Components, Certified by The Japan Petroleum Institute, TCI America
Catalog # TCS0429-SET
Supplier: TCI America
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Gasoline components
- Cat. No.:TCS0429-SET
- Contents:Methanol 1.5 vol% level; Ethanol 5.3 vol% level; MTBE 5.2 vol% level; ETBE 5.2 vol% level; Benzene 0.5 vol% level; Toluene 16.2 vol% level; Xylene 10.7 vol% level; Kerosene 3.7 vol% level
- Supplier no.:S0429-SET
Specifications
About this item
Certifications: Certified by The Japan Petroleum Institute