Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Phthalic anhydride ≥98.0%

Phthalic anhydride ≥98.0%

500 g
Catalog # TCP1614-500G
Supplier:   TCI America
Supplier #:  P1614-500G
Phthalic anhydride ≥98.0%
Catalog # TCP1614-500G
Supplier:  TCI America
CAS Number:  85-44-9
Phthalic anhydride ≥98.0%

500 g

CAS Number:  85-44-9
Price
$20.00
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 85-44-9
Molecular Formula: C8H4O3
Molecular Weight: 148.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 284
Melting point (°C): 131
Flash Point (°C): 152

  • CAS Number:85-44-9
  • Formula:C₈H₄O₃
  • MDL Number:MFCD00005918
  • Molecular Weight:148.12 g/mol
  • Boiling Pt:284 °C
  • Melting Pt:131 °C
  • Flash Pt:152 °C
  • UN:2214
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Purity/Analysis Method:
    >98.0% (GC,T)

Specifications

  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.