Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ethidium bromide ≥95.0%

Ethidium bromide ≥95.0%
Supplier:  TCI America
Certificates
Ethidium bromide ≥95.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 1239-45-8
MDL Number: MFCD00011724
Molecular Formula: C21H20BrN3
Molecular Weight: 394.32
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 100

Product Categories: Bioscience, Stains & Dyes, Fluorescent Stains, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium Bromide, Homidium Bromide

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Flash Pt:100 °C
  • UN:2811
  • ADR:6.1,I
Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    >95.0% (HPLC,T)
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $96.00
    $96.00
    Each (1g)
       $96.00

    Supplier Number: E0370-1G
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $454.00
    $454.00
    Each (5g)
       $454.00

    Supplier Number: E0370-5G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.