Order Entry
Puerto Rico
ContactUsLinkComponent
Diethyl ether, anhydrous ≥99.5% (by GC) stabilized
Diethyl ether, anhydrous ≥99.5% (by GC) stabilized
Catalog # TCD3479-500ML
Supplier:  TCI America
CAS Number:  
undefined
Diethyl ether, anhydrous ≥99.5% (by GC) stabilized
Catalog # TCD3479-500ML
Supplier:  TCI America
Supplier Number:  D3479-500ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 ml
  • Cat. No.:
    TCD3479-500ML
  • Supplier no.:
    D3479-500ML

Specifications

Specification Test Results

  • Purity/Analysis Method:
    >99.5% (GC)

Specification Test Results

About this item

CAS Number:60-29-7
 :
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
 :74.12 g/mol
Boiling Pt:35 °C
Melting Pt:–116 °C
Density:0.72 (20 °C)
Flash Pt:–45 °C
UN:1155
ADR:3,I