Order Entry
Puerto Rico
ContactUsLinkComponent
Boron trichloride ∼9% ∼1.0 mol/L in dichloromethane
Boron trichloride ∼9% ∼1.0 mol/L in dichloromethane
Catalog # TCB3556-100ML
Supplier:  TCI America
CAS Number:  
Boron trichloride ∼9% ∼1.0 mol/L in dichloromethane
Catalog # TCB3556-100ML
Supplier:  TCI America
Supplier Number:  B3556-100ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 ml
  • Cat. No.:
    TCB3556-100ML
  • Supplier no.:
    B3556-100ML

Specifications

About this item

 :
 :117.17
UN:1741
ADR:2,
CAS Number:10294-34-5
Formula:BCl₃
MDL Number:MFCD00011313