Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Recombinant IGF-I (from E. coli)
Rat Recombinant IGF-I (from E. coli)
Certificates
undefined
Rat Recombinant IGF-I (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P08025
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    mechano growth factor|IGF-IA|Somatomedin C
  • UniProtKB:
    P08025
  • Protein/Peptide Name:
    IGF-I
  • Purity:
    ≥95%
  • Molecular Weight:
    7.7 kDa
  • Sequence:
    GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

Related Information