Mouse Recombinant SDF-1beta (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Stromal cell-derived factor-1 beta (SDF-1 β), also called CXCL12b, is one of two SDF-1 splice variants made by a wide variety of cells upon stimulation by inflammatory cytokines such as TNF, IL-1, and LPS. SDF-1 β signals through the G protein-coupled receptor CXCR4 to recruit activated leukocytes.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P40224-2
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CXCL12|PBSF
- Protein/Peptide Name:SDF-1beta / CXCL12
- Purity:≥95%
- Molecular Weight:8.5 kDa
- Sequence:KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Shipping Temperature:RT
- Grade:RUO
Specifications
Frequently Bought Together