Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant IL-17E (from E. coli)
Mouse Recombinant IL-17E (from E. coli)
Certificates
undefined
Mouse Recombinant IL-17E (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q8VHC9
  • Reconstitution Instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    IL-25
  • Protein/Peptide Name:
    IL-17E
  • Purity:
    ≥90%
  • Molecular Weight:
    17.6/35.2 kDa
  • Sequence:
    MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

Related Information

Frequently Bought Together