About this item
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: HUVEC proliferation, ED50 ≤8 ng/ml, bioactivity ≥1.3×10⁵ units/mg
- Gene ID:P16612-2
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:glioma-derived endothelial cell mitogen|VEGF-A|VPF
- UniProtKB:P16612-2
- Protein/Peptide Name:VEGF-165
- Purity:≥95%
- Molecular Weight:19.4/38.7 kDa
- Sequence:MAPTTEGEQK AHEVVKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNVTMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPENHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Related Information
Literature reference - PubMed ID 24702044
Additional Product Details
Shenandoah_Biotechnology_Product_Catalog