Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant MIP-1 A (from E. coli)
Mouse Recombinant MIP-1 A (from E. coli)
Certificates
undefined
Mouse Recombinant MIP-1 A (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: THP-1 chemotaxis, ED50 ≤100 ng/ml, bioactivity ≥NA
  • Gene ID:
    Q5QNW0
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    CCL3|LD78α
  • UniProtKB:
    Q5QNW0
  • Protein/Peptide Name:
    MIP-1 A
  • Purity:
    ≥95%
  • Molecular Weight:
    7.9 kDa
  • Sequence:
    APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

Related Information

Frequently Bought Together