Mouse Recombinant MCP-3 (from E. coli)
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q03366
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:MARC|CCL7
- Protein/Peptide Name:MCP-3
- Purity:≥95%
- Molecular Weight:8.5 kDa
- Sequence:QPDGPNASTC CYVKKQKIPK RNLKSYRRIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTP TPKP
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Frequently Bought Together